Learn More
Waters Corp Waters OBD Prep Column, C18, 5 μm, 19 mm x 100 mm
Supplier: Waters Corp 186002567
SunFire C18 columns are designed for general purpose method development, particularly at low pH. Featuring a highly retentive C18 ligand, these columns are well-suited for the purification of basic compounds and for conducting impurity profile assays.
- C18 chemistry
- Reversed phase separation
- Silica particle substrate
- pH range: 2 to 8
- Max. temperature: 45 °C
- Max. pressure: 6000 psi (415 Bar)
- Endcapped
- Low silanol activity
- Particle size: 5 μm
- Pore size: 100 Å
- Format: OBD Prep Column
- System: HPLC Preparative
- Technique: LC/MS, LC
- Inner diameter: 19 mm
- Length: 100 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.