Learn More
Waters Corp Waters OBD Prep Column, C18, 100Å, 10 μm, 19 x 150 mm
Supplier: Waters Corp 186002668
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds, and is ideally suited for purification and impurity profile assays.
- Chemistry: C18
- Separation mode: Reversed phase
- Particle substrate: Silica
- pH range: 2 to 8 pH
- Temperature limits: 45 °C
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped: Yes
- Silanol activity: Low
- Particle shape: Spherical
- Particle size: 10 μm
- Pore size: 100 Å
- Surface area: 340
- System: HPLC Preparative
- Technique: LC/MS, LC
- USP classification: L1
- Inner diameter: 19 mm
- Length: 150 mm
- Carbon load: 16 %
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.