Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Preparative Column, C18, 100Å, 30 x 50 mm

Catalog No. 50465994 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive for basic compounds, and is ideally suited for purification and impurity profile assays.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8 pH
  • Temperature limits: 45 °C
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: 100 Å
  • Format: OBD prep column
  • System: HPLC preparative
  • Technique: LC/MS, LC
  • USP classification: L1
  • Inner diameter: 30 mm
  • Length: 50 mm
  • Carbon load: 16%

Catalog No. 50-465-994 Supplier Waters Corp Supplier No. 186003854
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive for basic compounds, and is ideally suited for purification and impurity profile assays.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8 pH
  • Temperature limits: 45 °C
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: 100 Å
  • Format: OBD prep column
  • System: HPLC preparative
  • Technique: LC/MS, LC
  • USP classification: L1
  • Inner diameter: 30 mm
  • Length: 50 mm
  • Carbon load: 16%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.