Learn More
Waters Corp SunFire C18 OBD Prep Column, 100Å, 10 µm, 30 mm X 250 mm, 1/pk
Catalog No. 50781275 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.
Catalog No. 50-781-275 Supplier Waters Corp Supplier No. 186002671
Add to Cart
Add to Cart
Request a Quote
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.