Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters OBD Prep Column, C18, 10 μm, 50 mm × 250 mm

Catalog No. 50781277 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.

  • C18 chemistry.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Temperature limits up to 45 °C.
  • Maximum pressure of 6000 psi (415 bar).
  • Endcapped column.
  • Low silanol activity.
  • Spherical particle shape.
  • Particle size of 10 μm.
  • Pore size of 100 Å.
  • OBD prep column format.
  • Designed for HPLC preparative systems.
  • Suitable for LC/MS and LC techniques.
  • Inner diameter of 50 mm.
  • Length of 250 mm.
  • Carbon load of 16%.

Catalog No. 50-781-277 Supplier Waters Corp Supplier No. 186002673
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.

  • C18 chemistry.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Temperature limits up to 45 °C.
  • Maximum pressure of 6000 psi (415 bar).
  • Endcapped column.
  • Low silanol activity.
  • Spherical particle shape.
  • Particle size of 10 μm.
  • Pore size of 100 Å.
  • OBD prep column format.
  • Designed for HPLC preparative systems.
  • Suitable for LC/MS and LC techniques.
  • Inner diameter of 50 mm.
  • Length of 250 mm.
  • Carbon load of 16%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.