Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters SunFire OBD Prep Column, C18, 10 μm, 100 Å

Catalog No. 50782099 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C18 columns are used for general purpose method development at low pH. Their C18 ligand is highly retentive for basic compounds and ideally suited for purification and impurity profile assays.

  • Designed for low pH method development and purification.
  • C18 ligand offers high retention for basic compounds.
  • Provides uniform, optimally packed bed density with direct scalability.
  • Ensures excellent column lifetime and consistent performance.
  • Delivers good resolution, peak shape, and high mass loading.
  • Maximizes recoveries and reduces risk in Prep method development.
  • pH range: 2 to 8.
  • Temperature limit: 45 °C.
  • Maximum pressure: 6000 psi (415 bar).
  • Particle size: 10 μm.
  • Pore size: 100 Å.
  • Dimensions: 30 mm inner diameter, 100 mm length.

Catalog No. 50-782-099 Supplier Waters Corp Supplier No. 186003971
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

SunFire C18 columns are used for general purpose method development at low pH. Their C18 ligand is highly retentive for basic compounds and ideally suited for purification and impurity profile assays.

  • Designed for low pH method development and purification.
  • C18 ligand offers high retention for basic compounds.
  • Provides uniform, optimally packed bed density with direct scalability.
  • Ensures excellent column lifetime and consistent performance.
  • Delivers good resolution, peak shape, and high mass loading.
  • Maximizes recoveries and reduces risk in Prep method development.
  • pH range: 2 to 8.
  • Temperature limit: 45 °C.
  • Maximum pressure: 6000 psi (415 bar).
  • Particle size: 10 μm.
  • Pore size: 100 Å.
  • Dimensions: 30 mm inner diameter, 100 mm length.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.