Learn More
Waters Corp Waters Corporation Preparative Column, C18, 10 μm, 50 mm x 100 mm, 100Å
Catalog No. 50782100 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These preparative columns are designed for general purpose method development at low pH. Their C18 ligand provides high retention, particularly for basic compounds, making them ideally suited for purification and impurity profile assays.
  • General purpose method development at low pH.
  • Highly retentive C18 ligand for basic compounds.
  • Ideal for purification and impurity profile assays.
  • Reversed phase separation.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Temperature limit of 45°C.
  • Maximum pressure of 6000 psi (415 Bar).
  • 10 μm spherical particles.
  • 100 Å pore size.
  • 50 mm inner diameter and 100 mm length.
  • 16% carbon load.
Catalog No. 50-782-100 Supplier Waters Corp Supplier No. 186003972
Add to Cart
Add to Cart
Request a Quote
These preparative columns are designed for general purpose method development at low pH. Their C18 ligand provides high retention, particularly for basic compounds, making them ideally suited for purification and impurity profile assays.
  • General purpose method development at low pH.
  • Highly retentive C18 ligand for basic compounds.
  • Ideal for purification and impurity profile assays.
  • Reversed phase separation.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Temperature limit of 45°C.
  • Maximum pressure of 6000 psi (415 Bar).
  • 10 μm spherical particles.
  • 100 Å pore size.
  • 50 mm inner diameter and 100 mm length.
  • 16% carbon load.