Learn More
Waters Corp Waters OBD Prep Column, C18, 10 μm, 100 Å, 30 mm x 150 mm
Catalog No. 50781274 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are for general purpose low-pH method development. The highly retentive C18 ligand suits purification and impurity profile assays. This OBD Prep Column provides consistent results in preparative chromatography, with its Optimum Bed Density (OBD) design eliminating method re-development.
  • Used for general-purpose low-pH method development.
  • C18 ligand provides high retention, ideal for purification and impurity profile assays.
  • Optimum Bed Density (OBD) design eliminates method re-development.
  • Manufactured in a cGMP-compliant facility.
  • Bonded silica structure ensures high-mass loading and low pH stability.
  • Engineered distributors and inert seals ensure positive chromatographic separations.
  • Increased mechanical stability is vital for laboratory-scale HPLC purification.
  • Delivers reliable and predictable data.
Catalog No. 50-781-274 Supplier Waters Corp Supplier No. 186002670
Add to Cart
Add to Cart
Request a Quote
SunFire C18 columns are for general purpose low-pH method development. The highly retentive C18 ligand suits purification and impurity profile assays. This OBD Prep Column provides consistent results in preparative chromatography, with its Optimum Bed Density (OBD) design eliminating method re-development.
  • Used for general-purpose low-pH method development.
  • C18 ligand provides high retention, ideal for purification and impurity profile assays.
  • Optimum Bed Density (OBD) design eliminates method re-development.
  • Manufactured in a cGMP-compliant facility.
  • Bonded silica structure ensures high-mass loading and low pH stability.
  • Engineered distributors and inert seals ensure positive chromatographic separations.
  • Increased mechanical stability is vital for laboratory-scale HPLC purification.
  • Delivers reliable and predictable data.