Learn More
Waters Corp Preparative Column, C18, OBD, 10 μm, 19 x 50 mm
Supplier:  Waters Corp 186002667
Encompass_Preferred
The SunFire Preparative Column is designed for high-performance preparative chromatography. Utilizing OBD Technology, it ensures a uniform and optimally packed bed density, providing direct scalability from analytical to preparative applications.
  • Utilizes OBD Technology
  • Features uniform, optimum packed bed density
  • Provides direct scalability from analytical to preparative chromatography
  • Offers excellent column lifetime
  • Ensures consistent column-to-column performance
  • Delivers good resolution and peak shape
  • Allows for high mass loading for unparalleled preparative chromatography results
Catalog No. 50-781-271