Learn More
Waters Corp Waters OBD Prep Column, C18, 100 Å, 10 μm, 19 mm x 250 mm
Catalog No. 50781273 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters SunFire C18 OBD Prep Column is a preparative HPLC column with C18 chemistry, ideal for method development and purification at low pH. It offers high stability and retention, integrating an Optimum Bed Density design for superior chromatographic performance in mass spectrometry applications.
  • Greater low pH stability.
  • Optimum Bed Density (OBD) column design.
  • Advanced silica synthesis and bonding technologies.
  • Improved peak shape performance.
  • Withstands pressures up to 6000 psi (415 Bar).
  • Highly retentive for basic compounds.
  • Achieves sharp peaks, good sensitivity, and low bleed in mass spectrometry.
  • Offers outstanding resolution, speed, and low back-pressure.
Catalog No. 50-781-273 Supplier Waters Corp Supplier No. 186002669
Add to Cart
Add to Cart
The Waters SunFire C18 OBD Prep Column is a preparative HPLC column with C18 chemistry, ideal for method development and purification at low pH. It offers high stability and retention, integrating an Optimum Bed Density design for superior chromatographic performance in mass spectrometry applications.
  • Greater low pH stability.
  • Optimum Bed Density (OBD) column design.
  • Advanced silica synthesis and bonding technologies.
  • Improved peak shape performance.
  • Withstands pressures up to 6000 psi (415 Bar).
  • Highly retentive for basic compounds.
  • Achieves sharp peaks, good sensitivity, and low bleed in mass spectrometry.
  • Offers outstanding resolution, speed, and low back-pressure.