Learn More
Waters Corp Waters C18 OBD Prep Column, 100Å, 5μm, 30 mm × 75 mm
Supplier: Waters Corp 186002571
C18 columns are designed for general purpose method development at low pH. They feature a highly retentive C18 ligand, making them particularly suitable for basic compounds. These columns are ideally suited for purification and impurity profile assays.
- High retention: the C18 ligand offers high retention, especially for basic compounds.
- Low pH method development: suitable for general purpose method development at low pH.
- Purification and impurity profiling: ideally suited for purification and impurity profile assays.
- Low silanol activity.
- The column is endcapped.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.