Learn More
Waters Corp Waters C18 OBD Prep Column, 100Å, 5 μm, 30 mm x 250 mm
Catalog No. 50782097 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are designed for low pH method development, providing high retention for basic compounds. Ideal for purification and impurity profile assays, they offer excellent low-pH stability and high chromatographic efficiency. Proprietary technologies ensure superior peak shapes and greater sensitivity, enabling seamless scale-up from analytical to preparative applications.
  • Excellent low-pH stability.
  • High chromatographic efficiency.
  • Superior peak shapes for charged analytes.
  • Symmetrical peaks for improved resolution and quantitation.
  • Greater sensitivity.
  • OBD Column design for equivalent performance during scale-up.
Catalog No. 50-782-097 Supplier Waters Corp Supplier No. 186003969
Add to Cart
Add to Cart
Request a Quote
SunFire C18 columns are designed for low pH method development, providing high retention for basic compounds. Ideal for purification and impurity profile assays, they offer excellent low-pH stability and high chromatographic efficiency. Proprietary technologies ensure superior peak shapes and greater sensitivity, enabling seamless scale-up from analytical to preparative applications.
  • Excellent low-pH stability.
  • High chromatographic efficiency.
  • Superior peak shapes for charged analytes.
  • Symmetrical peaks for improved resolution and quantitation.
  • Greater sensitivity.
  • OBD Column design for equivalent performance during scale-up.