Learn More
Waters Corp Waters SunFire C18 OBD Prep Column, 5 μm Particle Size, 100 Å Pore Size, 30 mm × 100 mm
Catalog No. 50781212 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are designed for general purpose method development at low pH. They feature a highly retentive C18 ligand, which is particularly effective for basic compounds, making them suitable for purification and impurity profile assays.
  • Highly retentive for basic compounds
  • Ideally suited for purification and impurity profile assays
  • Designed for general purpose method development at low pH
  • Operates within a pH range of 2 to 8
  • Withstands maximum pressure of 6000 psi (415 Bar)
Catalog No. 50-781-212 Supplier Waters Corp Supplier No. 186002572
Add to Cart
Add to Cart
Request a Quote
SunFire C18 columns are designed for general purpose method development at low pH. They feature a highly retentive C18 ligand, which is particularly effective for basic compounds, making them suitable for purification and impurity profile assays.
  • Highly retentive for basic compounds
  • Ideally suited for purification and impurity profile assays
  • Designed for general purpose method development at low pH
  • Operates within a pH range of 2 to 8
  • Withstands maximum pressure of 6000 psi (415 Bar)