Learn More
Waters Corp Waters SunFire C18 OBD Prep Column, 100Å, 5 μm, 19 × 50 mm
Supplier: Waters Corp 186002566
SunFire C18 columns are designed for general purpose method development in low pH environments, featuring a highly retentive C18 ligand. They are particularly suitable for basic compounds, purification, and impurity profile assays.
- Designed for general purpose method development in low pH conditions.
- Highly retentive C18 ligand for basic compounds.
- Suitable for purification and impurity profile assays.
- Chemistry: C18
- Separation Mode: Reversed Phase
- Particle Substrate: Silica
- Particle Size: 5 μm
- Pore Size: 100 Å
- Inner Diameter: 19 mm
- Length: 50 mm
- Format: OBD Prep Column
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.