Learn More
Waters Corp Waters OBD Prep Column, C18, 100Å Pore, 5 μm Particle, 50 mm x 100 mm
Supplier: Waters Corp 186002869
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds, and is ideally suited for purification and impurity profile assays.
- Separation mode: reversed phase.
- Particle substrate: silica.
- pH range: 2 to 8.
- Maximum operating temperature: 45 °C.
- Maximum pressure: 6000 psi (415 bar).
- Endcapped and features low silanol activity.
- Spherical particle shape with 5 μm size and 100 Å pore size.
- Surface area: 340.
- Designed for HPLC preparative systems, LC/MS, and LC techniques.
- USP classification: L1.
- Dimensions: 50 mm inner diameter and 100 mm length.
- Carbon load: 16 %.
- Units per package: 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.