Learn More
Waters Corp Waters SunFire OBD Prep Column, C18, 5 µm, 50 mm x 250 mm
Catalog No. 50782098 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C18 columns are designed for general purpose method development at low pH. Their C18 ligand is highly retentive, especially for basic compounds, making them ideally suited for purification and impurity profile assays.
  • Utilizes patented optimum bed density design and packing process for uniform density.
  • Engineered to prevent over-compression and voids, ensuring mechanical stability.
  • Offers increased productivity with higher recoveries and longer column lifetimes.
  • Facilitates fast, efficient, lab-scale separations for greater throughput.
  • Provides direct scalability from UPLC, UHPLC, or HPLC for purification.
  • Ensures consistent column-to-column performance and extended lifetimes.
  • Features high mass loading capabilities.
Catalog No. 50-782-098 Supplier Waters Corp Supplier No. 186003970
Add to Cart
Add to Cart
Request a Quote
These C18 columns are designed for general purpose method development at low pH. Their C18 ligand is highly retentive, especially for basic compounds, making them ideally suited for purification and impurity profile assays.
  • Utilizes patented optimum bed density design and packing process for uniform density.
  • Engineered to prevent over-compression and voids, ensuring mechanical stability.
  • Offers increased productivity with higher recoveries and longer column lifetimes.
  • Facilitates fast, efficient, lab-scale separations for greater throughput.
  • Provides direct scalability from UPLC, UHPLC, or HPLC for purification.
  • Ensures consistent column-to-column performance and extended lifetimes.
  • Features high mass loading capabilities.