Learn More
Waters Corp Waters SunFire C18 Column, 100 Å, 5 μm, 19 x 150 mm
Supplier:  Waters Corp 186002568
Encompass_Preferred
These preparative HPLC columns are designed for general purpose method development at low pH. Featuring a highly retentive C18 ligand, they are ideally suited for purification and impurity profile assays. The Optimum Bed Density (OBD) design ensures consistent chromatographic results and aids in method re-development.
  • Optimized Bed Density (OBD) design ensures consistent results.
  • Highly retentive C18 ligand, especially for basic compounds.
  • Suitable for purification and impurity profile assays.
  • Bonded-silica architecture provides high-mass loading capability.
  • Remarkable low pH stability.
  • Specially engineered distributors and chemically inert seals ensure successful separations.
  • cGMP-compliant manufacturing for precise scale-up.
  • Increased mechanical stability for laboratory-scale HPLC purification.
Catalog No. 50-781-208