Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters SunFire C8 Guard Cartridge, 10 μm, 100 Å, 19 mm × 10 mm

Catalog No. 50781352 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C8 columns are designed for general-purpose method development at low pH. The C8 ligand is less hydrophobic and, consequently, less retentive compared to SunFire C18 columns, making it suitable for specific assays.

  • C8 chemistry for reversed-phase separation
  • Silica particle substrate
  • pH range of 2 to 8
  • Temperature limit of 45 °C
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped for improved performance
  • Low silanol activity
  • Spherical particle shape
  • Particle size of 10 μm
  • Pore size of 100 Å
  • Preparative guard cartridge format
  • Surface area of 340
  • Compatible with HPLC systems
  • Suitable for LC/MS and LC techniques
  • USP classification L7
  • Internal diameter of 19 mm
  • Length of 10 mm
  • Carbon load of 12%
  • Supplied as 1 unit per package

Catalog No. 50-781-352 Supplier Waters Corp Supplier No. 186002761
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SunFire C8 columns are designed for general-purpose method development at low pH. The C8 ligand is less hydrophobic and, consequently, less retentive compared to SunFire C18 columns, making it suitable for specific assays.

  • C8 chemistry for reversed-phase separation
  • Silica particle substrate
  • pH range of 2 to 8
  • Temperature limit of 45 °C
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped for improved performance
  • Low silanol activity
  • Spherical particle shape
  • Particle size of 10 μm
  • Pore size of 100 Å
  • Preparative guard cartridge format
  • Surface area of 340
  • Compatible with HPLC systems
  • Suitable for LC/MS and LC techniques
  • USP classification L7
  • Internal diameter of 19 mm
  • Length of 10 mm
  • Carbon load of 12%
  • Supplied as 1 unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.