Learn More
Waters Corp SunFire Prep Guard Cartridge, C8, 100Å, 5 μm, 19 x 10 mm
Catalog No. 50781345 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The SunFire C8 Prep Guard Cartridge is designed to protect prep columns from impurities, extending column lifespan and enhancing analytical performance. Featuring a 100Å, 5μm sorbent bed on a silica substrate, it efficiently retains particulates and contaminants while allowing target analytes to pass. It is compatible with UPLC systems and suitable for various applications including protein purification and metabolomics.
  • Retains particulates and contaminants with a 100Å, 5μm sorbent bed.
  • Ensures durable performance via bonded silica substrate.
  • Guarantees compatibility and easy integration into lab setups.
  • Extends prep column longevity and operational efficiency.
  • Enhances UPLC system analytical performance.
  • Easy to install and incorporate into workflow.
  • Offers cost-effective, single-use solution for column protection.
Catalog No. 50-781-345 Supplier Waters Corp Supplier No. 186002754
The SunFire C8 Prep Guard Cartridge is designed to protect prep columns from impurities, extending column lifespan and enhancing analytical performance. Featuring a 100Å, 5μm sorbent bed on a silica substrate, it efficiently retains particulates and contaminants while allowing target analytes to pass. It is compatible with UPLC systems and suitable for various applications including protein purification and metabolomics.
  • Retains particulates and contaminants with a 100Å, 5μm sorbent bed.
  • Ensures durable performance via bonded silica substrate.
  • Guarantees compatibility and easy integration into lab setups.
  • Extends prep column longevity and operational efficiency.
  • Enhances UPLC system analytical performance.
  • Easy to install and incorporate into workflow.
  • Offers cost-effective, single-use solution for column protection.