Learn More
Waters Corp SunFire C8 OBD Prep Column, 10 μm, 150 mm
Catalog No. 50781356 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These SunFire C8 columns are ideal for general purpose method development at low pH. Featuring a C8 ligand, they are less hydrophobic and less retentive than C18, making them suitable for specific assays. OBD technology ensures optimum packed bed density, allowing direct scalability from analytical to preparative liquid chromatography (LC).
  • General purpose method development at low pH.
  • C8 ligand offers less hydrophobicity and retention than C18.
  • Uniform, optimum packed bed density.
  • Direct scalability from analytical to preparative LC.
  • Excellent column lifetime.
  • Consistent column-to-column performance.
  • Good resolution, peak shape, and mass loading for preparative LC.
  • Isolation of targeted small molecules and biomolecules.
  • Traditional stainless steel column technology.
Catalog No. 50-781-356 Supplier Waters Corp Supplier No. 186002765
Add to Cart
Add to Cart
Request a Quote
These SunFire C8 columns are ideal for general purpose method development at low pH. Featuring a C8 ligand, they are less hydrophobic and less retentive than C18, making them suitable for specific assays. OBD technology ensures optimum packed bed density, allowing direct scalability from analytical to preparative liquid chromatography (LC).
  • General purpose method development at low pH.
  • C8 ligand offers less hydrophobicity and retention than C18.
  • Uniform, optimum packed bed density.
  • Direct scalability from analytical to preparative LC.
  • Excellent column lifetime.
  • Consistent column-to-column performance.
  • Good resolution, peak shape, and mass loading for preparative LC.
  • Isolation of targeted small molecules and biomolecules.
  • Traditional stainless steel column technology.