Learn More
Waters Corp Waters Preparative Column, C8, OBD Column, 10 μm, 19 x 150 mm
Catalog No. 50781354 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
OBD Preparative Columns optimize Liquid Chromatography (Prep LC) by ensuring uniform, optimally packed beds for direct scalability from analytical to preparative applications. They offer extended column lifetime and consistent performance, contributing to superior resolution, peak shape, and mass loading for reliable purification results.
  • Uniform, optimally packed beds for ideal preparative outcomes.
  • Exceptional column lifetimes ensure durability and long-term use.
  • Uncompromised scale-up from UPLC™ to preparative chromatography.
  • Maximized recoveries with robust packings for isolating molecules.
  • Reduced method development risk simplifies the process.
  • Consistent performance provides reliable, reproducible results.
  • Enhanced chromatographic performance: good resolution, peak shape, and mass loading.
Catalog No. 50-781-354 Supplier Waters Corp Supplier No. 186002763
OBD Preparative Columns optimize Liquid Chromatography (Prep LC) by ensuring uniform, optimally packed beds for direct scalability from analytical to preparative applications. They offer extended column lifetime and consistent performance, contributing to superior resolution, peak shape, and mass loading for reliable purification results.
  • Uniform, optimally packed beds for ideal preparative outcomes.
  • Exceptional column lifetimes ensure durability and long-term use.
  • Uncompromised scale-up from UPLC™ to preparative chromatography.
  • Maximized recoveries with robust packings for isolating molecules.
  • Reduced method development risk simplifies the process.
  • Consistent performance provides reliable, reproducible results.
  • Enhanced chromatographic performance: good resolution, peak shape, and mass loading.