Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters SunFire Prep Column, C8, 100Å 10 μm, 30 × 250 mm

Catalog No. 50781357 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C8 columns are designed for general purpose method development at low pH. The C8 ligand offers less hydrophobicity and retentivity compared to C18, making it suitable for specific assays. These OBD (Optimum Bed Density) prep columns ensure robust performance.

  • C8 chemistry for general purpose method development.
  • Reversed phase with silica substrate.
  • pH range: 2 to 8.
  • Temperature limit: 45 °C.
  • Max pressure: 6000 psi (415 Bar).
  • Endcapped with low silanol activity.
  • Spherical particle, 10 μm size.
  • 100 Å pore size; 340 surface area.
  • For HPLC preparative systems; LC/MS, LC techniques.
  • L7 USP classification.
  • Dimensions: 30 mm ID, 250 mm length.
  • 12% carbon load.
  • 1 unit per package.

Catalog No. 50-781-357 Supplier Waters Corp Supplier No. 186002766
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

SunFire C8 columns are designed for general purpose method development at low pH. The C8 ligand offers less hydrophobicity and retentivity compared to C18, making it suitable for specific assays. These OBD (Optimum Bed Density) prep columns ensure robust performance.

  • C8 chemistry for general purpose method development.
  • Reversed phase with silica substrate.
  • pH range: 2 to 8.
  • Temperature limit: 45 °C.
  • Max pressure: 6000 psi (415 Bar).
  • Endcapped with low silanol activity.
  • Spherical particle, 10 μm size.
  • 100 Å pore size; 340 surface area.
  • For HPLC preparative systems; LC/MS, LC techniques.
  • L7 USP classification.
  • Dimensions: 30 mm ID, 250 mm length.
  • 12% carbon load.
  • 1 unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.