Learn More
Waters Corp Waters OBD Prep Column, C8, 100 Å, 5 μm, 50 × 50 mm
Supplier: Waters Corp 186002868
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18.
- C8 chemistry for specific selectivity.
- Designed for reversed phase separation mode.
- Silica particle substrate.
- pH range of 2 to 8 pH.
- Operates up to 45 °C.
- Withstands maximum pressure of 6000 psi (415 bar).
- Endcapped for optimal performance.
- Features low silanol activity.
- Spherical particle shape for consistent separations.
- Particle size of 5 μm.
- Pore size of 100 Å.
- OBD preparative column format.
- Suitable for HPLC preparative systems.
- Supports LC/MS and LC techniques.
- Dimensions: 50 mm inner diameter and 50 mm length.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.