Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters OBD Preparative Column, C8, 5 μm, 19 mm x 150 mm

Catalog No. 50781344 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This OBD preparative column utilizes C8 chemistry, ideal for general purpose method development, particularly at low pH. Its C8 ligand offers less hydrophobicity and retention compared to C18 columns, making it suitable for specific assay requirements.

  • Separation mode: Reversed-Phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limit: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped design
  • Low silanol activity
  • Spherical particle shape
  • Particle size: 5 μm
  • Pore diameter: 100 Å
  • Format: OBD Preparative Column
  • Surface area: 340
  • System compatibility: HPLC Preparative
  • Techniques supported: LC/MS, LC
  • USP classification: L7
  • Inner diameter: 19 mm
  • Length: 150 mm
  • Carbon load: 12%
  • One unit per package

Catalog No. 50-781-344 Supplier Waters Corp Supplier No. 186002753
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This OBD preparative column utilizes C8 chemistry, ideal for general purpose method development, particularly at low pH. Its C8 ligand offers less hydrophobicity and retention compared to C18 columns, making it suitable for specific assay requirements.

  • Separation mode: Reversed-Phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limit: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped design
  • Low silanol activity
  • Spherical particle shape
  • Particle size: 5 μm
  • Pore diameter: 100 Å
  • Format: OBD Preparative Column
  • Surface area: 340
  • System compatibility: HPLC Preparative
  • Techniques supported: LC/MS, LC
  • USP classification: L7
  • Inner diameter: 19 mm
  • Length: 150 mm
  • Carbon load: 12%
  • One unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.