Learn More
Waters Corp Waters OBD Prep Column, C8, 100Å, 5 μm, 50 mm X 100 mm
Catalog No. 50781413 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
  • Used for general purpose method development at low pH.
  • C8 ligand is less hydrophobic and less retentive compared to C18 columns, making it preferred for some assays.
Catalog No. 50-781-413 Supplier Waters Corp Supplier No. 186002870
Add to Cart
Add to Cart
Request a Quote
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
  • Used for general purpose method development at low pH.
  • C8 ligand is less hydrophobic and less retentive compared to C18 columns, making it preferred for some assays.