Learn More
Waters Corp Waters OBD Prep Column, C8, 100Å, 5 μm, 30 mm × 75 mm
Catalog No. 50781347 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
  • General purpose method development at low pH
  • Less hydrophobic and less retentive compared to C18
Catalog No. 50-781-347 Supplier Waters Corp Supplier No. 186002756
Add to Cart
Add to Cart
Request a Quote
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
  • General purpose method development at low pH
  • Less hydrophobic and less retentive compared to C18