Learn More
Waters Corp SunFire Silica Prep Guard Cartridge, 100Å, 10 µm, 19 mm X 10 mm, 1/pk
Catalog No. 50781777 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire columns represent a new era in HPLC column performance and technology. The state-of-the art reversed-phase C18 and C8 bonded silica delivers excellent peak shape, high efficiency, low MS bleed and excellent low pH stability as well as high mass loading capability. SunFire columns are ideally suited for demanding HPLC applications where high-performance, reproducible and reliable columns are required. The Optimum Bed Density (OBD) designed preparative column performance coupled with the choice of C18, C8 and Normal-Phase Silica chemistries makes SunFire an excellent choice for successful analytical-to-lab scale purification.
Catalog No. 50-781-777 Supplier Waters Corp Supplier No. 186003444
SunFire columns represent a new era in HPLC column performance and technology. The state-of-the art reversed-phase C18 and C8 bonded silica delivers excellent peak shape, high efficiency, low MS bleed and excellent low pH stability as well as high mass loading capability. SunFire columns are ideally suited for demanding HPLC applications where high-performance, reproducible and reliable columns are required. The Optimum Bed Density (OBD) designed preparative column performance coupled with the choice of C18, C8 and Normal-Phase Silica chemistries makes SunFire an excellent choice for successful analytical-to-lab scale purification.