Learn More
Waters Corp SunFire Silica OBD Prep Column, 10 μm, 19 x 250 mm, 100Å
Supplier:  Waters Corp 186003447
Encompass_Preferred
Waters SunFire Silica OBD Prep Columns utilize Optimum Bed Density (OBD™) technology for high-performance lab-scale separations, ensuring consistent results, extended column lifetimes, and seamless scalability from analytical to preparative HPLC methods. This robust design optimizes productivity and reduces operational costs.
  • Achieve fast, efficient lab-scale separations.
  • Directly scale analytical to preparative HPLC methods.
  • Utilize robust chromatographic particles for purification.
  • Patented OBD design ensures uniform bed density, preventing voids.
  • Exceptional resistance to mechanical chromatographic bed failure.
  • Deliver consistent column-to-column performance, extending lifetimes.
  • Maximize productivity in preparative HPLC applications.
  • Silica chemistry with 100Å pore size and 10 μm particle size.
Catalog No. 50-781-780