Learn More
Waters Corp Waters SunFire Preparative Column, Silica, 50 x 50 mm, 10 μm, 100 Å
Catalog No. 50781783 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SunFire Preparative Column is designed to bridge the performance gap in Prep LC, offering uniform, optimum packed bed density for high-quality preparative chromatography. It features a Silica chemistry type, with dimensions of 50 x 50 mm, a particle size of 10 μm, and a pore size of 100 Å.
  • Exceptional column lifetimes.
  • Uncompromised scale-up from UPLC™ to Prep.
  • Maximized recoveries with robust packings.
  • Decreased risk of additional Prep method development.
  • Consistent column to column performance.
Catalog No. 50-781-783 Supplier Waters Corp Supplier No. 186003450
Add to Cart
Add to Cart
Request a Quote
This SunFire Preparative Column is designed to bridge the performance gap in Prep LC, offering uniform, optimum packed bed density for high-quality preparative chromatography. It features a Silica chemistry type, with dimensions of 50 x 50 mm, a particle size of 10 μm, and a pore size of 100 Å.
  • Exceptional column lifetimes.
  • Uncompromised scale-up from UPLC™ to Prep.
  • Maximized recoveries with robust packings.
  • Decreased risk of additional Prep method development.
  • Consistent column to column performance.