Learn More
Waters Corp Waters Preparative Column, Silica, 5 μm, 100 Å, 30 mm x 150 mm
Catalog No. 50781771 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preparative column utilizes traditional stainless steel OBD hardware and is considered a gold-standard in lab-scale preparative liquid chromatography.
  • Exceptional column lifetimes.
  • Uncompromised scale-up from UPLC™ to Prep.
  • Maximized recoveries with robust packings designed for isolation of targeted small molecules and biomolecules.
  • Decreased risk of additional preparative method development.
Catalog No. 50-781-771 Supplier Waters Corp Supplier No. 186003438
Add to Cart
Add to Cart
Request a Quote
This preparative column utilizes traditional stainless steel OBD hardware and is considered a gold-standard in lab-scale preparative liquid chromatography.
  • Exceptional column lifetimes.
  • Uncompromised scale-up from UPLC™ to Prep.
  • Maximized recoveries with robust packings designed for isolation of targeted small molecules and biomolecules.
  • Decreased risk of additional preparative method development.