Learn More
Waters Corp Waters Preparative Column, Silica, 5 μm, 19 x 100 mm
Catalog No. 50781765 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire Preparative Columns feature patented Optimum Bed Density (OBD) technology for fast, efficient lab-scale separations. They enable direct scaling from UPLC, UHPLC, or HPLC screening to purification, employing robust chromatographic particles. The OBD design ensures uniform density profiles, resists mechanical failure, and delivers consistent performance with extended lifetimes.
  • Fast, efficient lab-scale separations.
  • Direct scaling from UPLC, UHPLC, or HPLC to lab-scale purification.
  • Robust chromatographic particles for purification.
  • Patented OBD design ensures uniform density profiles, preventing voids.
  • Exceptional resistance to mechanical chromatographic bed failure.
  • Consistent column performance and extended lifetimes.
  • Maximizes productivity when scaling up screening methods.
  • Provides high mass loading.
Catalog No. 50-781-765 Supplier Waters Corp Supplier No. 186003432
Add to Cart
Add to Cart
SunFire Preparative Columns feature patented Optimum Bed Density (OBD) technology for fast, efficient lab-scale separations. They enable direct scaling from UPLC, UHPLC, or HPLC screening to purification, employing robust chromatographic particles. The OBD design ensures uniform density profiles, resists mechanical failure, and delivers consistent performance with extended lifetimes.
  • Fast, efficient lab-scale separations.
  • Direct scaling from UPLC, UHPLC, or HPLC to lab-scale purification.
  • Robust chromatographic particles for purification.
  • Patented OBD design ensures uniform density profiles, preventing voids.
  • Exceptional resistance to mechanical chromatographic bed failure.
  • Consistent column performance and extended lifetimes.
  • Maximizes productivity when scaling up screening methods.
  • Provides high mass loading.