Learn More
Waters Corp Waters SunFire Silica OBD Column, 100 Å, 5 μm, 50 x 100 mm
Catalog No. 50781773 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire Silica OBD Preparative Columns are designed for high mass loading applications. They are part of the Optimum Bed Density (OBD) Preparative Columns, which aim to increase productivity through higher recoveries and longer column lifetimes.
  • OBD Preparative Columns provide predictable, uniform density profiles throughout the column due to their patented design and packing process, which eliminates the potential for voids.
  • They exhibit exceptional resistance to mechanical chromatographic bed failure and deliver consistent column-to-column performance, extending lifetimes and reducing costs.
  • Silica columns are characterized by high mass loading capabilities.
Catalog No. 50-781-773 Supplier Waters Corp Supplier No. 186003440
Add to Cart
Add to Cart
Request a Quote
SunFire Silica OBD Preparative Columns are designed for high mass loading applications. They are part of the Optimum Bed Density (OBD) Preparative Columns, which aim to increase productivity through higher recoveries and longer column lifetimes.
  • OBD Preparative Columns provide predictable, uniform density profiles throughout the column due to their patented design and packing process, which eliminates the potential for voids.
  • They exhibit exceptional resistance to mechanical chromatographic bed failure and deliver consistent column-to-column performance, extending lifetimes and reducing costs.
  • Silica columns are characterized by high mass loading capabilities.