Learn More
Waters Corp SunFire Silica OBD Prep Column, 10 μm, 19 x 50 mm
Catalog No. 50781778 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire Prep OBD Silica columns are preparative columns used under normal phase conditions. The Optimum Bed Density (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
  • Preparative column
  • Normal phase conditions
  • Optimum Bed Density (OBD) hardware format
  • Superior column lifetime
  • Helps bridge the performance gap in preparative liquid chromatography (Prep LC)
  • Offers direct scalability from analytical to preparative
  • Ensures consistent column-to-column performance
  • Provides good resolution, peak shape, and mass loading for preparative chromatography
  • Realizes exceptional column lifetimes
  • Achieves uncompromised scale-up from UPLC™ to preparative
  • Maximizes recoveries with robust packings
  • Decreases risk of additional preparative method development
Catalog No. 50-781-778 Supplier Waters Corp Supplier No. 186003445
SunFire Prep OBD Silica columns are preparative columns used under normal phase conditions. The Optimum Bed Density (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
  • Preparative column
  • Normal phase conditions
  • Optimum Bed Density (OBD) hardware format
  • Superior column lifetime
  • Helps bridge the performance gap in preparative liquid chromatography (Prep LC)
  • Offers direct scalability from analytical to preparative
  • Ensures consistent column-to-column performance
  • Provides good resolution, peak shape, and mass loading for preparative chromatography
  • Realizes exceptional column lifetimes
  • Achieves uncompromised scale-up from UPLC™ to preparative
  • Maximizes recoveries with robust packings
  • Decreases risk of additional preparative method development