Learn More
Waters Corp HPLC Guard Cartridge, Silica C8, 100 Å, 10 μm, 30 x 10 mm
Catalog No. 502392842 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC guard cartridge is designed for high-performance liquid chromatography (HPLC) applications. It protects analytical and preparative columns from chemical and particulate contaminants, ensuring extended column lifetime and consistent chromatographic performance.
  • Stationary phase is C8 silica
  • Pore size of 100 Å
  • Particle size of 10 μm
  • Dimensions are 30 mm x 10 mm
Catalog No. 50-239-2842 Supplier Waters Corp Supplier No. 186006886
This HPLC guard cartridge is designed for high-performance liquid chromatography (HPLC) applications. It protects analytical and preparative columns from chemical and particulate contaminants, ensuring extended column lifetime and consistent chromatographic performance.
  • Stationary phase is C8 silica
  • Pore size of 100 Å
  • Particle size of 10 μm
  • Dimensions are 30 mm x 10 mm