Learn More
Waters Corp Preparative Column, Silica, 5 μm, 10 x 100 mm
Catalog No. 502392321 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preparative column utilizes OBD technology and silica chemistry to bridge the performance gap in preparative liquid chromatography. It offers uniform, optimum packed bed density and direct scalability from analytical to preparative applications. The design ensures excellent column lifetime and consistent performance for high-quality chromatography.
  • Exceptional column lifetimes.
  • Uncompromised scale-up from analytical to preparative applications.
  • Maximized recoveries with robust packings for isolating targeted small molecules and biomolecules.
  • Decreased risk of additional preparative method development.
  • Provides good resolution, peak shape, and mass loading for high-quality preparative chromatography.
Catalog No. 50-239-2321 Supplier Waters Corp Supplier No. 186008181
This preparative column utilizes OBD technology and silica chemistry to bridge the performance gap in preparative liquid chromatography. It offers uniform, optimum packed bed density and direct scalability from analytical to preparative applications. The design ensures excellent column lifetime and consistent performance for high-quality chromatography.
  • Exceptional column lifetimes.
  • Uncompromised scale-up from analytical to preparative applications.
  • Maximized recoveries with robust packings for isolating targeted small molecules and biomolecules.
  • Decreased risk of additional preparative method development.
  • Provides good resolution, peak shape, and mass loading for high-quality preparative chromatography.