Learn More
Waters Corp OBD Prep Column, Silica, 5 μm, 10 mm, 250 mm, 100 Å
Supplier: Waters Corp 186008183
SunFire Prep Silica columns are general purpose columns designed for use under normal phase conditions. This product contains hazardous material, which requires special freight handling, and additional charges may apply.
- Chemistry: Silica
- Separation mode: Normal-phase
- Particle substrate: Silica
- pH range: 2 to 8 pH
- Temperature limits: 45 °C
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped: No
- Bonding technology: Silica
- Silanol activity: High
- Particle shape: Spherical
- Particle size: 5 μm
- Pore size: 100 Å
- Format: OBD Prep Column
- Surface area: 340
- System: HPLC Preparative
- Technique: LC/MS, LC
- USP classification: L3
- Inner diameter: 10 mm
- Length: 250 mm
- Product type: Preparative columns
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.