Learn More
Waters Corp SunFire Prep Guard Cartridge, 100Å, 5 μm, 30 mm ID, 10 mm Length
Catalog No. 502392730 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Prep Guard Cartridge is designed to protect both HPLC and SFC 30 mm ID Prep OBD™ columns. When placed before the prep column, this guard assembly helps extend column lifetimes and reduce purification costs.
  • Designed to protect both HPLC and SFC 30 mm ID Prep OBD™ columns.
  • Extends column lifetimes.
  • Reduces purification costs.
Catalog No. 50-239-2730 Supplier Waters Corp Supplier No. 186006889
This Prep Guard Cartridge is designed to protect both HPLC and SFC 30 mm ID Prep OBD™ columns. When placed before the prep column, this guard assembly helps extend column lifetimes and reduce purification costs.
  • Designed to protect both HPLC and SFC 30 mm ID Prep OBD™ columns.
  • Extends column lifetimes.
  • Reduces purification costs.