Learn More
Waters Corp Waters OBD Prep Column, C18, 5 μm, 10 mm x 50 mm
Supplier:  Waters Corp 186008152
Encompass_Preferred
C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds, and it is ideally suited for purification and impurity profile assays.
  • General purpose method development at low pH.
  • Highly retentive C18 ligand.
  • Ideally suited for purification and impurity profile assays.
Catalog No. 50-138-4639