Learn More
Sigma Aldrich Fine Chemicals Biosciences Sunflower seed oil from He1L
Catalog No. 502902664
Encompass
Sunflower seed oil from Helianthus annuus contains saturated fatty acids (palmitic and stearic acids) monounsaturated fatty acid (oleic acid) and polyunsaturated acid (mostly linoleic acid).1
Catalog No. 50-290-2664 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S50071L
Sunflower seed oil from Helianthus annuus contains saturated fatty acids (palmitic and stearic acids) monounsaturated fatty acid (oleic acid) and polyunsaturated acid (mostly linoleic acid).1