Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sunflower Seed Oil, Spectrum™ Chemical
SDP

Catalog No. 18607191 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
200 L
Packaging:
Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-191 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-191 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1929200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1929, 8001-21-6

Spectrum™ Chemical Sunflower Seed Oil is, as its name suggests, the oil of sunflower seeds that is extracted by the compression of the seeds. This non-volatile oil is most commonly used in food and as a frying oil. It is also used in cosmetics as an emollient. It is also mixed with diesel to run diesel combustion engines.

Chemical Identifiers

CAS 8001-21-6

Specifications

CAS Max % 1
Packaging Drum
Quantity 200 L
Saponification Value 188-194
Specific Gravity 0.914-0.924
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Sunflower Seed Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.