Learn More
AUTOMATIONDIRECT COM INC SUNG-IL MCHNRY DRV CPLNG 6MM
Catalog No. NC3066810
Encompass
Sung-Il Machinery drive coupling hub, jaw type, aluminum alloy,clamp, size 20, 6mm bore. Requires SJC-20 series spiders.
Catalog No. NC3066810 Supplier AUTOMATIONDIRECT COM INC Supplier No. SJC20C6
Sung-Il Machinery drive coupling hub, jaw type, aluminum alloy,clamp, size 20, 6mm bore. Requires SJC-20 series spiders.