Learn More
AUTOMATIONDIRECT COM INC SUNG-IL MCHNRY DRV CPLNG SPDR
Catalog No. NC3066686
Encompass
Sung-Il Machinery drive coupling spider, jaw type, size 20,thermoplastic polyurethane, 98A durometer. For use with SJC-20Cseries hubs.
Catalog No. NC3066686 Supplier AUTOMATIONDIRECT COM INC Supplier No. SJC20BLSLEEVE
Sung-Il Machinery drive coupling spider, jaw type, size 20,thermoplastic polyurethane, 98A durometer. For use with SJC-20Cseries hubs.