Learn More
Apexbio Technology LLC Sunitinib 557795-19-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062630
Encompass_Preferred
Sunitinib (CAS 557795-19-4) is an orally bioavailable small molecule inhibitor targeting multiple receptor tyrosine kinases (RTKs) including VEGFR1-3 PDGFR / c-kit and RET Research has shown sunitinib inhibits proliferation in nasopharyngeal carcinoma (NPC) cell lines HK1 CNE-1 CNE-2 HONE-1 and C666-1 with IC50 values ranging from approximately 2 06 to 7 57 M Additionally it induces apoptosis and G0/G1 cell cycle arrest in NPC cell lines and significantly decreases microvessel density in CNE-2 xenograft tumor models in vivo Thus sunitinib is useful for investigating pathways involved in tumor angiogenesis and proliferation
Catalog No. 50-000-62630 Supplier Apexbio Technology LLC Supplier No. B10455.1
Sunitinib (CAS 557795-19-4) is an orally bioavailable small molecule inhibitor targeting multiple receptor tyrosine kinases (RTKs) including VEGFR1-3 PDGFR / c-kit and RET Research has shown sunitinib inhibits proliferation in nasopharyngeal carcinoma (NPC) cell lines HK1 CNE-1 CNE-2 HONE-1 and C666-1 with IC50 values ranging from approximately 2 06 to 7 57 M Additionally it induces apoptosis and G0/G1 cell cycle arrest in NPC cell lines and significantly decreases microvessel density in CNE-2 xenograft tumor models in vivo Thus sunitinib is useful for investigating pathways involved in tumor angiogenesis and proliferation