Learn More
Medchemexpress LLC HY-10255A 500mg , Sunitinib CAS:557795-19-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10255A500MG
Encompass_Preferred
HY-10255A 500mg Sunitinib CAS: 557795-19-4 Sunitinib (SU 11248) is a multi-targeted receptor tyrosine kinase inhibitor with IC50s of 80 nM and 2 nM for VEGFR2 and PDGFRβ, respectively. Sunitinib, an ATP-competitive inhibitor, effectively inhibits autophosphorylation of Ire1α by inhibiting autophosphorylation and consequent RNase activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9038