Learn More
Selleck Chemical LLC Sunitinib
SDP
Catalog No. 5000001463
Encompass_Preferred
Sunitinib is a multi-targeted RTK inhibitor targeting VEGFR2 (Flk-1) and PDGFR with IC50 of 80 nM and 2 nM and also inhibits c-Kit Sunitinib is also a dose-dependent inhibitor of the autophosphorylation activity of IRE1 Sunitinib induces autophagy and apoptosis
Catalog No. 50-000-01463 Supplier Selleck Chemical LLC Supplier No. S77811G
Sunitinib is a multi-targeted RTK inhibitor targeting VEGFR2 (Flk-1) and PDGFR with IC50 of 80 nM and 2 nM and also inhibits c-Kit Sunitinib is also a dose-dependent inhibitor of the autophosphorylation activity of IRE1 Sunitinib induces autophagy and apoptosis