Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sunitinib-d10 | 1126721-82-1 | MFCD09841296 | 99.9% | 408.54 g/mol | C22H17D10FN4O2 | 1 MG
SDP

Catalog No. 5000246340
Encompass_Preferred

Sunitinib-d10 is a deuterium-labeled form of the receptor tyrosine kinase inhibitor sunitinib, supplied as a solid powder intended for use as an isotope-labeled internal standard or tracer in quantitative bioanalytical workflows (LC-MS, GC-MS, NMR). It is provided in small analytical package sizes and requires low-temperature storage for long-term stability.

  • Deuterium-labeled (d10) internal standard for quantitative mass spectrometry and tracer studies.
  • High reported purity (99.89%) suitable for analytical applications.
  • Chemical formula C22H17D10FN4O2 and molecular weight 408.54 g/mol.
  • Available in low-mass packages for analytical use and stable when stored at recommended low temperatures.

Catalog No. 50-002-46340 Supplier Medchemexpress LLC Supplier No. HY10255AS1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sunitinib-d10 is a deuterium-labeled form of the receptor tyrosine kinase inhibitor sunitinib, supplied as a solid powder intended for use as an isotope-labeled internal standard or tracer in quantitative bioanalytical workflows (LC-MS, GC-MS, NMR). It is provided in small analytical package sizes and requires low-temperature storage for long-term stability.

  • Deuterium-labeled (d10) internal standard for quantitative mass spectrometry and tracer studies.
  • High reported purity (99.89%) suitable for analytical applications.
  • Chemical formula C22H17D10FN4O2 and molecular weight 408.54 g/mol.
  • Available in low-mass packages for analytical use and stable when stored at recommended low temperatures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.