Learn More
Abcam Sunitinib, Free base, 100MG
Catalog No. 502671245
Encompass_Preferred
MW 398.5 Da, Purity >=99%. Sunitinib is a free base form of Sunitinib malate. A CSF-1 receptor kinase inhibitor .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1245 Supplier Abcam Supplier No. AB285423100MG
MW 398.5 Da, Purity >=99%. Sunitinib is a free base form of Sunitinib malate. A CSF-1 receptor kinase inhibitor .
The product is subject to the following: Abcam Restricted Use Statement