Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sunitinib impurity 28 | 1186651-51-3 | 98.5% | 299.31 | 50 MG
SDP

Catalog No. 5000401637
Encompass_Preferred

Sunitinib impurity 28 is a well-characterized impurity of Sunitinib, identified as an important reference standard. It is provided with high purity and a defined molecular weight, making it suitable for research and analytical applications.

  • Purity: 98.5%
  • Molecular weight: 299.31
  • Formula: C16H14FN3O2
  • CAS number: 1186651-51-3
  • Appearance: Solid
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-004-01637 Supplier Medchemexpress LLC Supplier No. HY4485650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sunitinib impurity 28 is a well-characterized impurity of Sunitinib, identified as an important reference standard. It is provided with high purity and a defined molecular weight, making it suitable for research and analytical applications.

  • Purity: 98.5%
  • Molecular weight: 299.31
  • Formula: C16H14FN3O2
  • CAS number: 1186651-51-3
  • Appearance: Solid
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.