Learn More
Selleck Chemical LLC Sunitinib malate S1042-10mM/1mL
SDP
Catalog No. 503108465
Encompass_Preferred
Sunitinib malate is a multi-targeted RTK inhibitor targeting VEGFR2 (Flk-1) and PDGFR with IC50 of 80 nM and 2 nM in cell-free assays and also inhibits c-Kit Sunitinib Malate effectively inhibits autophosphorylation of Ire1 Sunitinib Malate increases both death receptor and mitochondrial-dependent apoptosis
Catalog No. 50-310-8465 Supplier Selleck Chemical LLC Supplier No. S104210MM/1ML
Sunitinib malate is a multi-targeted RTK inhibitor targeting VEGFR2 (Flk-1) and PDGFR with IC50 of 80 nM and 2 nM in cell-free assays and also inhibits c-Kit Sunitinib Malate effectively inhibits autophosphorylation of Ire1 Sunitinib Malate increases both death receptor and mitochondrial-dependent apoptosis