Learn More
Apexbio Technology LLC Sunitinib malate(Synonyms: Sutent, SU11248, Sunitinib L-malate, Sunitinib malate salt, SU-11248 malate, Sutent malate), 2g, CAS: 341031-54-7.
SDP
Catalog No. 5000061050
Encompass_Preferred
Sunitinib malate (CAS 341031-54-7) is a small molecule orally bioavailable multitargeted receptor tyrosine kinase inhibitor It inhibits various receptor tyrosine kinases including platelet-derived growth factor receptor (PDGFR) vascular endothelial growth factor receptors (VEGFR-1 2 and 3) c-KIT FLT3 kinase colony-stimulating factor-1 receptor (CSF-1R) and RET kinase In neuroblastoma cell lines (e g SK-N-BE(2) SH-SY5Y LAN-5) treatment with sunitinib malate significantly suppresses proliferation in a dose-dependent manner (IC50 10-20 ng/ml) Preclinical studies show tumor growth inhibition and reduced metastasis in neuroblastoma mouse models highlighting its potential for research in oncology therapeutics
Catalog No. 50-000-61050 Supplier Apexbio Technology LLC Supplier No. A82552000
Sunitinib malate (CAS 341031-54-7) is a small molecule orally bioavailable multitargeted receptor tyrosine kinase inhibitor It inhibits various receptor tyrosine kinases including platelet-derived growth factor receptor (PDGFR) vascular endothelial growth factor receptors (VEGFR-1 2 and 3) c-KIT FLT3 kinase colony-stimulating factor-1 receptor (CSF-1R) and RET kinase In neuroblastoma cell lines (e g SK-N-BE(2) SH-SY5Y LAN-5) treatment with sunitinib malate significantly suppresses proliferation in a dose-dependent manner (IC50 10-20 ng/ml) Preclinical studies show tumor growth inhibition and reduced metastasis in neuroblastoma mouse models highlighting its potential for research in oncology therapeutics